Learn More
Sino Biological Recombinant Human EGFR Protein (T790M, C797S, L858R, GST Tag), Active
Catalog No. 502575205
Encompass_Preferred
Recombinant human EGFR (T790M C797S L858R) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5205 Supplier Sino Biological Supplier No. E10122VG50
Recombinant human EGFR (T790M C797S L858R) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.