Learn More
Sino Biological Recombinant Human EGFR Protein (d746-750 T790M L792H, GST Tag), Active
Catalog No. 502575269
Encompass_Preferred
Recombinant human EGFR (d746-750 T790M L792H) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5269 Supplier Sino Biological Supplier No. E10122F3G50
Recombinant human EGFR (d746-750 T790M L792H) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.