Learn More
Sino Biological Recombinant Human EGFR Protein (d746-750 T790M L792F, GST Tag), Active
Catalog No. 502575264
Encompass_Preferred
Recombinant human EGFR (d746-750 T790M L792F) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5264 Supplier Sino Biological Supplier No. E10122G3G500
Recombinant human EGFR (d746-750 T790M L792F) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.