Learn More
Eurofins Discoverx EGFR (L858R), active, 250 µg. (14-626M)
Supplier:  Eurofins Discoverx 14626M
Encompass
N-terminal GST-tagged recombinant, human EGF receptor amino acids 696-end containing the mutation L858R. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9080