Learn More
Eurofins Discoverx EGFR (L861Q), active, 250 µg. (14-627M)
Supplier:  Eurofins Discoverx 14627M
Encompass
N-terminal GST-tagged recombinant, human EGF receptor amino acids 696-end containing the mutation L861Q. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9082