Learn More
Medchemexpress LLC EGFR Mouse HEK293 100ug
SDP
Catalog No. 5000180875
Encompass_Preferred
The EGFR protein is a receptor tyrosine kinase that binds EGF family ligands and activates multiple signaling cascades Ligand binding induces receptor homo- or heterodimerization leading to autophosphorylation and recruitment of adapter proteins (such as GRB2) EGFR Protein Mouse (HEK293 Fc) is the recombinant mouse-derived EGFR protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80875 Supplier Medchemexpress LLC Supplier No. HYP72988100UG
The EGFR protein is a receptor tyrosine kinase that binds EGF family ligands and activates multiple signaling cascades Ligand binding induces receptor homo- or heterodimerization leading to autophosphorylation and recruitment of adapter proteins (such as GRB2) EGFR Protein Mouse (HEK293 Fc) is the recombinant mouse-derived EGFR protein expressed by HEK293 with C-hFc labeled tag