Learn More
Medchemexpress LLC EGFR Mouse HEK293 10ug
Supplier: Medchemexpress LLC HYP7298810UG
The EGFR protein is a receptor tyrosine kinase that binds EGF family ligands and activates multiple signaling cascades Ligand binding induces receptor homo- or heterodimerization leading to autophosphorylation and recruitment of adapter proteins (such as GRB2) EGFR Protein Mouse (HEK293 Fc) is the recombinant mouse-derived EGFR protein expressed by HEK293 with C-hFc labeled tag