Learn More
Sino Biological Recombinant Human EGFR Protein (d746-750, C797A, GST Tag), Active
Catalog No. 502575289
Encompass_Preferred
Recombinant human EGFR (d746-750 C797A) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5289 Supplier Sino Biological Supplier No. E10122C2G50
Recombinant human EGFR (d746-750 C797A) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.