Learn More
ECM Bioscience EGFR Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience EK6160
Encompass_Preferred
The EGFR phospho-regulation antibody sampler kit can be used to detect EGFR phosphorylation at Ser-1142, Ser-967, and Tyr-1101. The kit also includes an antibody to examine total EGFR expression levels, and secondary reagents for rabbit polyclonal and mouse monoclonal antibody detection. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8694