Learn More
Sino Biological Recombinant Human EGFR Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10001H27HB100
Encompass_Preferred
A DNA sequence encoding the human EGFR (NP_005219.2) (Met1-Ser645) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5264