Learn More
Medchemexpress LLC EGFR Rat HEK293 H 500ug
SDP
Catalog No. 5000180760
Encompass_Preferred
EGFR Protein is a receptor tyrosine kinase that binds to ligands of the EGF family EGFR Protein is involved in the regulation of NF-kappa-B RAS-RAF-MEK-ERK PI3 kinase-AKT PLCgamma-PKC and STATs signaling pathways EGFR Protein has calmodulin binding activity epidermal growth factor binding activity and epidermal growth factor activating receptor activity EGFR Protein Rat (HEK293 His) is the recombinant rat-derived EGFR protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80760 Supplier Medchemexpress LLC Supplier No. HYP72990500UG
EGFR Protein is a receptor tyrosine kinase that binds to ligands of the EGF family EGFR Protein is involved in the regulation of NF-kappa-B RAS-RAF-MEK-ERK PI3 kinase-AKT PLCgamma-PKC and STATs signaling pathways EGFR Protein has calmodulin binding activity epidermal growth factor binding activity and epidermal growth factor activating receptor activity EGFR Protein Rat (HEK293 His) is the recombinant rat-derived EGFR protein expressed by HEK293 with C-His labeled tag