Learn More
Enzo Life Sciences EGF receptor (human) Recombinant monoclonal antibody (528) (Mouse IgG2a kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4820200
Encompass_Preferred
Clone: 528. Isotype: IgG2aκ. Immunogen: Partially purified EGFR from A431 cells. Species reactivity: Human. Applications: Flow Cytometry, IF, IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3009