Learn More
Medchemexpress LLC Egfr T790m/L858r-In-2 | 2955607-40-4 | 99.7% | 497.57 | 1 ML
SDP
Catalog No. 5000361714
Encompass_Preferred
EGFR T790M/L858R-IN-2 is a potent and selective inhibitor of EGFRT790M/L858R, with IC50 values of 3.5 nM for EGFRT790M/L858R and 1290 nM for EGFR WT. It reduces the expression of p-EGFR, P-AKT, and P-ERK1/2, and induces apoptosis and cell cycle arrest in the G1 phase. This compound demonstrates anti-cancer activity. It is intended for research use only and is not sold to patients.
  • Potent and selective EGFRT790M/L858R inhibitor
  • Reduces expression of p-EGFR, P-AKT, and P-ERK1/2
  • Induces apoptosis and cell cycle arrest in G1 phase
  • Demonstrates anti-cancer activity
Catalog No. 50-003-61714 Supplier Medchemexpress LLC Supplier No. HY14982410MM/1ML
EGFR T790M/L858R-IN-2 is a potent and selective inhibitor of EGFRT790M/L858R, with IC50 values of 3.5 nM for EGFRT790M/L858R and 1290 nM for EGFR WT. It reduces the expression of p-EGFR, P-AKT, and P-ERK1/2, and induces apoptosis and cell cycle arrest in the G1 phase. This compound demonstrates anti-cancer activity. It is intended for research use only and is not sold to patients.
  • Potent and selective EGFRT790M/L858R inhibitor
  • Reduces expression of p-EGFR, P-AKT, and P-ERK1/2
  • Induces apoptosis and cell cycle arrest in G1 phase
  • Demonstrates anti-cancer activity