Learn More
Medchemexpress LLC EGFR T790M/L858R-IN-2 | 2955607-40-4 | 99.7% | 497.57 | 100 MG
Supplier: Medchemexpress LLC HY149824100MG
EGFR T790M/L858R-IN-2 is a potent and selective inhibitor of EGFRT790M/L858R and EGFR WT. It decreases the expression of p-EGFR, P-AKT, and P-ERK1/2, and induces apoptosis and cell cycle arrest in the G1 phase, demonstrating anti-cancer activity.
- Potent and selective EGFRT790M/L858R inhibitor
- Decreases expression of p-EGFR, P-AKT, and P-ERK1/2
- Induces apoptosis
- Induces cell cycle arrest in the G1 phase
- Shows anti-cancer activity
- For research use only