Learn More
Medchemexpress LLC EGFR T790M/L858R-IN-2 | 2955607-40-4 | 99.7% | 497.57 | 25 MG
SDP
Catalog No. 5000360761
Encompass_Preferred
EGFR T790M/L858R-IN-2 is a potent and selective inhibitor of EGFRT790M/L858R and EGFR WT. It decreases the expression of p-EGFR, P-AKT, P-ERK1/2, induces apoptosis and cell cycle arrest in the G1 phase, and demonstrates anti-cancer activity.
  • Potent and selective EGFRT790M/L858R inhibitor
  • Decreases expression of p-EGFR, P-AKT, P-ERK1/2
  • Induces apoptosis and cell cycle arrest in G1 phase
  • Shows anti-cancer activity
Catalog No. 50-003-60761 Supplier Medchemexpress LLC Supplier No. 25MGHY149824
EGFR T790M/L858R-IN-2 is a potent and selective inhibitor of EGFRT790M/L858R and EGFR WT. It decreases the expression of p-EGFR, P-AKT, P-ERK1/2, induces apoptosis and cell cycle arrest in the G1 phase, and demonstrates anti-cancer activity.
  • Potent and selective EGFRT790M/L858R inhibitor
  • Decreases expression of p-EGFR, P-AKT, P-ERK1/2
  • Induces apoptosis and cell cycle arrest in G1 phase
  • Shows anti-cancer activity