Learn More
Medchemexpress LLC EGFR T790M/L858R-IN-8 | 333419-97-9 | 99.2% | 359.17 | 25 MG
SDP
Catalog No. 5000395458
Encompass_Preferred
EGFR T790M/L858R-IN-8 is a potent inhibitor of EGFR, specifically targeting the mutant EGFRT790M/L858R with an IC50 of 56.8 μM. While it effectively targets the mutant EGFR, its anti-proliferative effect on cancer cell lines A549, A431, and NHI-H1975 is not significant. This product is intended for research use only and is not for patient treatment.
  • Potent inhibitor of EGFR targeting mutant EGFRT790M/L858R
  • IC50: 56.8 μM
Catalog No. 50-003-95458 Supplier Medchemexpress LLC Supplier No. 25MGHY164056
EGFR T790M/L858R-IN-8 is a potent inhibitor of EGFR, specifically targeting the mutant EGFRT790M/L858R with an IC50 of 56.8 μM. While it effectively targets the mutant EGFR, its anti-proliferative effect on cancer cell lines A549, A431, and NHI-H1975 is not significant. This product is intended for research use only and is not for patient treatment.
  • Potent inhibitor of EGFR targeting mutant EGFRT790M/L858R
  • IC50: 56.8 μM