Learn More
Medchemexpress LLC EGFR vIII Human Bi 20ug | 20ug
SDP
Catalog No. 5000187570
Encompass_Preferred
The EGFRvIII protein is a transmembrane glycoprotein in the protein kinase superfamily that acts as a receptor for epidermal growth factor It acts on the cell surface binds to epidermal growth factor triggers receptor dimerization and tyrosine autophosphorylation and promotes cell proliferation EGFR vIII Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived EGFR vIII protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87570 Supplier Medchemexpress LLC Supplier No. HYP7811720UG
The EGFRvIII protein is a transmembrane glycoprotein in the protein kinase superfamily that acts as a receptor for epidermal growth factor It acts on the cell surface binds to epidermal growth factor triggers receptor dimerization and tyrosine autophosphorylation and promotes cell proliferation EGFR vIII Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived EGFR vIII protein expressed by HEK293 with C-Avi C-His labeled tag