Learn More
Cayman Chemical EGFR/ErbB2 Inhibitor, Purity: ≥95%, CAS: 179248-61-4, 5 mg
SDP
Catalog No. 502965755
Encompass_Preferred
A cell-permeable inhibitor of EGFR and c-ErbB2 (IC50s = 20 and 79 nM, respectively); inhibits the proliferation of cancer cells overexpressing either c-ErbB2 or EGFR (IC50s = 2.3-2.5 µM)
Catalog No. 50-296-5755 Supplier Cayman Chemical Supplier No. 175685 MG
A cell-permeable inhibitor of EGFR and c-ErbB2 (IC50s = 20 and 79 nM, respectively); inhibits the proliferation of cancer cells overexpressing either c-ErbB2 or EGFR (IC50s = 2.3-2.5 µM)