Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque EGF R Protein, Fc Tag;HEK293;100UG;EGR,EGFR,EGF R,ERBB,ERBB1,HER1,PIG61,mENA,EGF-R,ERBB-1,HER-1,EGFRvIII
Catalog No. 501658577
Encompass_Preferred
Recombinant Protein;Rhesus macaque EGF R Protein, Fc Tag;100UG;Rhesus macaque EGF R, Fc Tag (EGR-C5252) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P55245).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 95.3 kDa. The protein migrates as 130-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EGF, Tag Free (Cat. No. EGF-H4114) at 10 μg/mL (100 μL/well) can bind Rhesus macaque EGF R, Fc Tag (Cat. No. EGR-C5252) with a linear range of 39-625 μg/mL (QC tested).
Catalog No. 50-165-8577 Supplier ACROBiosystems Supplier No. EGRC5252100UG
Recombinant Protein;Rhesus macaque EGF R Protein, Fc Tag;100UG;Rhesus macaque EGF R, Fc Tag (EGR-C5252) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P55245).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 95.3 kDa. The protein migrates as 130-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EGF, Tag Free (Cat. No. EGF-H4114) at 10 μg/mL (100 μL/well) can bind Rhesus macaque EGF R, Fc Tag (Cat. No. EGR-C5252) with a linear range of 39-625 μg/mL (QC tested).