Learn More
ACROBiosystems Recombinant Protein;Human EGF R Protein, Fc Tag;HEK293;1MG;EGR,EGFR,EGF R,ERBB,ERBB1,HER1,PIG61,mENA,EGF-R,ERBB-1,HER-1,EGFRvIII
Catalog No. 501657556
Encompass_Preferred
Recombinant Protein;Human EGF R Protein, Fc Tag;1MG;Human EGF R, Fc Tag (EGR-H5252) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # NP_005219).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 95 kDa. The protein migrates as 130-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EGF, Tag Free (Cat. No. EGF-H4114) at 10μg/mL (100 μL/well) can bind Human EGF R, Fc Tag (Cat. No. EGR-H5252) with a linear range of 0.2-3.2 μg/mL (QC tested).;Immobilized Human EGF R, Fc Tag (Cat. No. EGR-H5252) at 1 μg/mL (100 μL/well) can bind Cetuximab with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-165-7556 Supplier ACROBiosystems Supplier No. EGRH52521MG
Recombinant Protein;Human EGF R Protein, Fc Tag;1MG;Human EGF R, Fc Tag (EGR-H5252) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # NP_005219).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 95 kDa. The protein migrates as 130-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EGF, Tag Free (Cat. No. EGF-H4114) at 10μg/mL (100 μL/well) can bind Human EGF R, Fc Tag (Cat. No. EGR-H5252) with a linear range of 0.2-3.2 μg/mL (QC tested).;Immobilized Human EGF R, Fc Tag (Cat. No. EGR-H5252) at 1 μg/mL (100 μL/well) can bind Cetuximab with a linear range of 0.1-2 ng/mL (Routinely tested).