Learn More
Cayman Chemical EGFRL858R RabbIt MoncLNal A
SDP
Catalog No. 502947126
Encompass_Preferred
A dual agonist of FXR and PPAR activates FXR and PPAR in reporter assays using HEK293T cells (EC50s 0.94 and 1.5 UM respectively) selective for these receptors over RARa RXRa PPARa PPARY and LXRa at 10 UM
Catalog No. 50-294-7126 Supplier Cayman Chemical Supplier No. 32327100 UL
A dual agonist of FXR and PPAR activates FXR and PPAR in reporter assays using HEK293T cells (EC50s 0.94 and 1.5 UM respectively) selective for these receptors over RARa RXRa PPARa PPARY and LXRa at 10 UM