Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human EGFRvIII Protein, His Tag
Catalog No. 501956449
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human EGFRvIII Protein, His Tag (EGI-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1). It is the FITC labeled form of Human EGFRvIII Protein, His Tag (EGI-H52H4).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa.
Catalog No. 50-195-6449 Supplier ACROBiosystems Supplier No. EGIHF2H7200UG
Recombinant Protein;200UG;FITC-Labeled Human EGFRvIII Protein, His Tag (EGI-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1). It is the FITC labeled form of Human EGFRvIII Protein, His Tag (EGI-H52H4).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa.