Learn More
ACROBiosystems ACROBiosystems Human EGFRvIII Protein, His Tag
Supplier:  ACROBiosystems EGIH52H41MG
Encompass_Preferred
Recombinant Protein;1MG;Human EGFRvIII, His Tag (EGI-H52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cetuximab at 2 μg/mL (100 μL/well) can bind Human EGFRvIII, His Tag (Cat. No. EGI-H52H4) with a linear range of 0.2-5 ng/mL (QC tested).
Catalog No. 50-195-7386