Learn More
CPC Scientific H-Ile-Ser-Ile-Asn-Gln-Asp-Leu-Lys-Ala-Ile-Thr-Asp-Met-Leu-Leu-Thr-Glu-Gln-Ile-Arg-Glu-Arg-Gln-Arg-Tyr-Leu-Ala-Asp-Leu-Arg-Gln-Arg-Leu-Leu-Glu-Lys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109217
Encompass
Egg-laying hormone (ELH) a neurotransmitter on neurons of the abdominal ganglion, act as hormone by diffusing to the circulating hemolymph to modulate the activity of other organs. It corresponds to aa 206-241 of ELH, and specifically causes contraction of smooth muscle in the ovotestis and expulsion of the egg string.ONE-LETTER SEQUENCE: ISINQDLKAITDMLLTEQIRERQRYLADLRQRLLEK-NH2MOLECULAR FORMULA: C190H329N59O57S1MOLECULAR WEIGHT:4384.13STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117680-39-4], RESEARCH AREA: HormonalREFERENCES: R.H.Scheller et al., Cell, 32, 7 (1983)
Catalog No. 50-210-9217 Supplier CPC Scientific Supplier No. ELH001A
Egg-laying hormone (ELH) a neurotransmitter on neurons of the abdominal ganglion, act as hormone by diffusing to the circulating hemolymph to modulate the activity of other organs. It corresponds to aa 206-241 of ELH, and specifically causes contraction of smooth muscle in the ovotestis and expulsion of the egg string.ONE-LETTER SEQUENCE: ISINQDLKAITDMLLTEQIRERQRYLADLRQRLLEK-NH2MOLECULAR FORMULA: C190H329N59O57S1MOLECULAR WEIGHT:4384.13STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117680-39-4], RESEARCH AREA: HormonalREFERENCES: R.H.Scheller et al., Cell, 32, 7 (1983)