Learn More
Medchemexpress LLC Egg oil | 8001-17-0 | 25 MG
SDP
Catalog No. 5000390156
Encompass_Preferred
Egg oil is a natural oil, which consists primarily of cholesterol, lecithin and glycerides of the fatty acids. Egg oil exhibits activity in regulating the gut microbial dysbiosis, alleviating obesity, insulin resistance and inflammation.
  • Reduces IL-8 secretion, demonstrating anti-inflammatory efficacy.
  • Protects against neurodegenerative diseases by attenuating cytotoxicity and modulating antioxidant enzymes.
  • Maintains cell viability in Caco2 cells.
  • Improves insulin sensitivity and hepatic glycogen synthesis, alleviating insulin resistance.
  • Ameliorates HFD-induced obesity, restoring gut microbial balance and down-regulating the LPS/TLR4 pathway.
  • Reduces body weight, adipose weight gain, blood glucose, and lipids in mice.
  • Increases mRNA levels of key signaling proteins in muscle and liver.
Catalog No. 50-003-90156 Supplier Medchemexpress LLC Supplier No. 25MGHYN12670
Egg oil is a natural oil, which consists primarily of cholesterol, lecithin and glycerides of the fatty acids. Egg oil exhibits activity in regulating the gut microbial dysbiosis, alleviating obesity, insulin resistance and inflammation.
  • Reduces IL-8 secretion, demonstrating anti-inflammatory efficacy.
  • Protects against neurodegenerative diseases by attenuating cytotoxicity and modulating antioxidant enzymes.
  • Maintains cell viability in Caco2 cells.
  • Improves insulin sensitivity and hepatic glycogen synthesis, alleviating insulin resistance.
  • Ameliorates HFD-induced obesity, restoring gut microbial balance and down-regulating the LPS/TLR4 pathway.
  • Reduces body weight, adipose weight gain, blood glucose, and lipids in mice.
  • Increases mRNA levels of key signaling proteins in muscle and liver.