Learn More
TARGETMOL CHEMICALS INC Egg PG 50MG
SDP
Catalog No. 502877273
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Egg PG(Phosphatidylglycerols(egg)) is one of the major phosphatidylglycerols of food and biological tissues, often found in the membranes of bacterial and animal cells, and serves as an anionic phospholipid substrate in the sPMCA, reaction. Purity 98%
Catalog No. 50-287-7273 Supplier TARGETMOL CHEMICALS INC Supplier No. TN724450MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Egg PG(Phosphatidylglycerols(egg)) is one of the major phosphatidylglycerols of food and biological tissues, often found in the membranes of bacterial and animal cells, and serves as an anionic phospholipid substrate in the sPMCA, reaction. Purity 98%