Learn More
Avanti Polar Lipids EGG PHOSPHATIDYLCHOLINE - 1G
Catalog No. NC9208084
Encompass
L-alpha-Phosphatidylcholine (Egg, Chicken), Hygroscopic as lyophilized powder, C 42 H 82 NO8P, Percent composition: C 66.37% H 10.87% N 1.84% O 16.84% P 4.08%, 1g
Catalog No. NC9208084 Supplier Avanti Polar Lipids Supplier No. 840051C1G
L-alpha-Phosphatidylcholine (Egg, Chicken), Hygroscopic as lyophilized powder, C 42 H 82 NO8P, Percent composition: C 66.37% H 10.87% N 1.84% O 16.84% P 4.08%, 1g