Learn More
Apexbio Technology LLC egg white lysozyme (19-36) [Gallus gallus], 10mg
SDP
Catalog No. 501904684
Encompass_Preferred
Sequence: H2N-KVFGRCELAAAMKRHGLD-OHFormula of Egg white lysozyme (19-36) [Gallus gallus]:C86H144N28O23S2Hen egg white lysozyme has a molecular weight of about 14,600 and each molecule comprises 129 amino acid residues1. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4684 Supplier Apexbio Technology LLC Supplier No. A106510
Sequence: H2N-KVFGRCELAAAMKRHGLD-OHFormula of Egg white lysozyme (19-36) [Gallus gallus]:C86H144N28O23S2Hen egg white lysozyme has a molecular weight of about 14,600 and each molecule comprises 129 amino acid residues1. Other sizes are available. Please inqury us for quote.