Learn More
NEOGEN EGG YOLK EMUL 50% 100ML 10PK
SDP
Supplier:  NEOGEN 700004878
Encompass_Preferred
Egg Yolk Emulsion 50% (100 mL) is a growth supplement intended for use in culture media for the microbial analysis of food, animal feed, water, and environmental samples. Sold per PK of 10 x 100ml.
Catalog No. NC2262655