Learn More
HIMEDIA Egg Yolk Tel Emulsion, FD046, Microbiology Culture Media, Enrichment Supplement, Egg Yolk Emulsion, Staphylococcus Identification, Sterile, Vials, 5 vials (100 ml per vial)
Catalog No. NC4013616
Encompass_Preferred
Egg Yolk Tel Emulsion is a sterile, stabilized potassium tellurite emulsion of egg yolk used to identify Staphylococcus species on selective media. Each 100 ml vial blends egg yolk, sterile saline and potassium tellurite. Once warmed to a uniform emulsion and added to molten base medium, it reveals lecithinase and lipase reactions and tellurite reduction that help differentiate staphylococci, including coagulase-positive strains. A ready-to-add enrichment for clinical and food microbiology. Store at 2 to 8 C. Packaged as 5 vials (100 ml per vial).
Catalog No. NC4013616 Supplier HIMEDIA Supplier No. FD046100MLX5VL
Egg Yolk Tel Emulsion is a sterile, stabilized potassium tellurite emulsion of egg yolk used to identify Staphylococcus species on selective media. Each 100 ml vial blends egg yolk, sterile saline and potassium tellurite. Once warmed to a uniform emulsion and added to molten base medium, it reveals lecithinase and lipase reactions and tellurite reduction that help differentiate staphylococci, including coagulase-positive strains. A ready-to-add enrichment for clinical and food microbiology. Store at 2 to 8 C. Packaged as 5 vials (100 ml per vial).