Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EGGGG-PEG8-amide-bis(deoxyglucitol) | 2842855-38-1 | 98.9% | C49H91N9O28 | 100 MG
SDP

Catalog No. 5000357536
Encompass_Preferred

EGGGG-PEG8-amide-bis(deoxyglucitol) is identified as a cleavable ADC (Antibody-Drug Conjugate) linker. It has a molecular weight of 1254.29 and appears as a solid-liquid mixture, white to off-white in color. For research use only, this product is not sold to patients. It is recommended to store the product at -20°C, protected from light. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light. Its solubility in DMSO is 75 mg/mL.

  • Product type: Cleavable ADC linker
  • Molecular weight: 1254.29
  • Appearance: Solid-liquid mixture
  • Color: White to off-white
  • Research use: For research only; not sold to patients
  • Storage (solid): -20°C, protected from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protected from light)
  • Solubility (in vitro): 75 mg/mL in DMSO

Catalog No. 50-003-57536 Supplier Medchemexpress LLC Supplier No. HY49484100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGGGG-PEG8-amide-bis(deoxyglucitol) is identified as a cleavable ADC (Antibody-Drug Conjugate) linker. It has a molecular weight of 1254.29 and appears as a solid-liquid mixture, white to off-white in color. For research use only, this product is not sold to patients. It is recommended to store the product at -20°C, protected from light. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light. Its solubility in DMSO is 75 mg/mL.

  • Product type: Cleavable ADC linker
  • Molecular weight: 1254.29
  • Appearance: Solid-liquid mixture
  • Color: White to off-white
  • Research use: For research only; not sold to patients
  • Storage (solid): -20°C, protected from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protected from light)
  • Solubility (in vitro): 75 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.