Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EGGGG-PEG8-amide-bis(deoxyglucitol) | 2842855-38-1 | MFCD27995527 | >98.9% | C49H91N9O28 | 25 MG
SDP

Catalog No. 5000358919
Encompass_Preferred

EGGGG-PEG8-amide-bis(deoxyglucitol) is a cleavable ADC linker. It is used in Antibody-drug Conjugate/ADC Related research.

  • Product type: ADC linker
  • Classification: Cleavable
  • Appearance: Solid-liquid mixture
  • Color: White to off-white
  • Intended use: For research use only; not for sale to patients

Catalog No. 50-003-58919 Supplier Medchemexpress LLC Supplier No. 25MGHY49484
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGGGG-PEG8-amide-bis(deoxyglucitol) is a cleavable ADC linker. It is used in Antibody-drug Conjugate/ADC Related research.

  • Product type: ADC linker
  • Classification: Cleavable
  • Appearance: Solid-liquid mixture
  • Color: White to off-white
  • Intended use: For research use only; not for sale to patients

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.