Learn More
Medchemexpress LLC Eggggg-peg8-amide-bis(deoxyglucitol) | 2842855-38-1 | 98.9% | 1254.29 g/mol | C49H91N9O28 | 10 MG
SDP
Catalog No. 5000298453
Encompass_Preferred
EGGGG-PEG8-amide-bis(deoxyglucitol) is a cleavable antibody-drug conjugate (ADC) linker for research applications. It provides a hydrophilic, cleavable spacer that combines PEG and deoxyglucitol moieties to facilitate conjugation, improve solubility, and enable controlled payload release.
  • High purity (98.9%).
  • Hydrophilic PEG-deoxyglucitol spacer enhances solubility.
  • Cleavable linker chemistry suitable for controlled payload release.
  • Soluble in DMSO at elevated temperature (75 mg/mL with warming).
  • Supplied as a white to off-white solid-liquid mixture for handling.
Catalog No. 50-002-98453 Supplier Medchemexpress LLC Supplier No. HY4948410MG
EGGGG-PEG8-amide-bis(deoxyglucitol) is a cleavable antibody-drug conjugate (ADC) linker for research applications. It provides a hydrophilic, cleavable spacer that combines PEG and deoxyglucitol moieties to facilitate conjugation, improve solubility, and enable controlled payload release.
  • High purity (98.9%).
  • Hydrophilic PEG-deoxyglucitol spacer enhances solubility.
  • Cleavable linker chemistry suitable for controlled payload release.
  • Soluble in DMSO at elevated temperature (75 mg/mL with warming).
  • Supplied as a white to off-white solid-liquid mixture for handling.