Learn More
Aviva Systems Biology Corp EGLN1 Antibody - C-terminal region (ARP34608_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34608_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EGLN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human EGLN1. Alternate names: HPH2, PHD2, SM20, ECYT3, HPH-2, HIFPH2, ZMYND6, C1orf12, HIF-PH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55478