Learn More
Aviva Systems Biology Corp EGLN2 Antibody - N-terminal region (ARP34747_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34747_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EGLN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EGLN2. Alternate names: EIT6, PHD1, HPH-1, HPH-3, HIFPH1, HIF-PH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-55544