Learn More
Aviva Systems Biology Corp Egln2 Antibody - C-terminal region (ARP39776_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39776_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Egln2. It was validated on Western Blot. . Alternate names: MGC93662, MGC93666, PHD-1, PHD1, HPH-1, HPH-3, HIF-PH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-827