Learn More
Lonza Walkersville EGM-Plus Endothelial Cell Growth Media-Plus BulletKit
Catalog No. NC0916308
Encompass
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Content:1 x EBM Plus Basal Medium (CC-5036), 475 mL1 x EGM Plus SingleQuots Supplement Pack (CC-4542) containing:1 x Bottle FBS, 10.00 mL1 x Vial Hydrocortisone, 0.50 mL1 x Bottle L-Glutamine, 25.00 mL1 x Vial EnGS, 1.00 mL1 x Vial Ascorbic Acid, 0.50 mL1 x Vial rhEGF, 0.50 mL1 x Vial GA-1000, 0.50 mL1 x Vial Heparin, 0.50 mL
Catalog No. NC0916308 Supplier Lonza Walkersville Supplier No. CC5035
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Content:1 x EBM Plus Basal Medium (CC-5036), 475 mL1 x EGM Plus SingleQuots Supplement Pack (CC-4542) containing:1 x Bottle FBS, 10.00 mL1 x Vial Hydrocortisone, 0.50 mL1 x Bottle L-Glutamine, 25.00 mL1 x Vial EnGS, 1.00 mL1 x Vial Ascorbic Acid, 0.50 mL1 x Vial rhEGF, 0.50 mL1 x Vial GA-1000, 0.50 mL1 x Vial Heparin, 0.50 mL