Learn More
Lonza Walkersville EGM Endothelial Cell Growth Medium SingleQuots Kit
Catalog No. NC9409994
Encompass
EGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the EGM Endothelial Cell Growth Medium BulletKit (Catalog Number CC-3124) which consists of basal medium and supplements packaged together.Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Content:1 x Green Cap Vial with hEGF 0.50 mL1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 Bottle BBE 2.00 mL1 x Blue Cap Vial with Ascorbic Acid 0.50 mL1 Bottle FBS 10.00 mL
Catalog No. NC9409994 Supplier Lonza Walkersville Supplier No. CC4133
EGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the EGM Endothelial Cell Growth Medium BulletKit (Catalog Number CC-3124) which consists of basal medium and supplements packaged together.Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Content:1 x Green Cap Vial with hEGF 0.50 mL1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 Bottle BBE 2.00 mL1 x Blue Cap Vial with Ascorbic Acid 0.50 mL1 Bottle FBS 10.00 mL