Learn More
Aviva Systems Biology Corp EGR1 Antibody - N-terminal region (ARP36841_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36841_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EGR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse EGR1. Alternate names: egr, TIS8, Zenk, Egr-1, NGFIA, Zfp-6, ETR103, Krox-1, Krox24, NGF1-A, NGFI-A, Zif268, Krox-24, A530045N19RikApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-56282