Learn More
Aviva Systems Biology Corp EGR1 Antibody - N-terminal region (ARP32241_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32241_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EGR1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EGR1. Alternate names: AT225, G0S30, KROX-24, NGFI-A, TIS8, ZIF-268, ZNF225ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54494