Learn More
Aviva Systems Biology Corp EGR2 Antibody - C-terminal region (P100880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EGR2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human EGR2. Alternate names: CMT1D, CMT4E, DKFZp686J1957, FLJ14547, KROX20, AT591ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62760