Learn More
Aviva Systems Biology Corp EGR3 Antibody - middle region (ARP34196_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34196_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EGR3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EGR3. Alternate names: MGC138484, PILOT, EGR-3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-63076