Learn More
Bioworld EGTA 0.5M, PH 8.0,250ML
Catalog No. NC1850504
Encompass_Preferred
EGTA 0.5M, pH 8.0, DNase and RNase Free,250ml.EGTA Buffer is of a 0.5M concentration and pH of 8.0 that is DNase and RNase free. EGTA Buffer is a metal chelating agent specific for calcium ions. EGTA Buffer is commonly used in tissue culture media and biological buffers.
Catalog No. NC1850504 Supplier Bioworld Supplier No. 401212663
EGTA 0.5M, pH 8.0, DNase and RNase Free,250ml.EGTA Buffer is of a 0.5M concentration and pH of 8.0 that is DNase and RNase free. EGTA Buffer is a metal chelating agent specific for calcium ions. EGTA Buffer is commonly used in tissue culture media and biological buffers.