Learn More
Bioworld EGTA 0.5M, pH 8.0, DNase and RNase Free, 50 mL
Catalog No. NC1874048
Encompass_Preferred
EGTA Buffer is of a 0.5M concentration and pH of 8.0 that is DNase and RNase free. EGTA Buffer is a metal chelating agent specific for calcium ions. EGTA Buffer is commonly used in tissue culture media and biological buffers.
Catalog No. NC1874048 Supplier Bioworld Supplier No. 401212661
EGTA Buffer is of a 0.5M concentration and pH of 8.0 that is DNase and RNase free. EGTA Buffer is a metal chelating agent specific for calcium ions. EGTA Buffer is commonly used in tissue culture media and biological buffers.