Learn More
Kd Medical Inc EGTA. 0.5M, PH8.0, 100ML BTL
SDP
Catalog No. 501031357
Encompass
EGTA is a chelating agent that can be used for the determination of calcium in the presence of magnesium. One 100ml bottle.
Catalog No. 50-103-1357 Supplier Kd Medical Inc Supplier No. PMB0702
EGTA is a chelating agent that can be used for the determination of calcium in the presence of magnesium. One 100ml bottle.