Learn More
Boston Bioproducts Inc EGTA (0.5 M, pH 8.0) - 50ml
Catalog No. NC9339264
Encompass_Preferred
EGTA solution, also known as ethylene glycol-bis(β-aminoethyl ether)-N,N,N’,N’-tetraacetic acidis extensively used in molecular biology and protein biochemistry applications that require the chelation of divalent metal cations, such as calcium ions. This specificity makes it particularly useful in biological systems where calcium plays an important role, as well as for the determination of calcium in the presence of magnesium, since EGTA has greater specificity for calcium ions.
Catalog No. NC9339264 Supplier Boston Bioproducts Inc Supplier No. BM15150ML
EGTA solution, also known as ethylene glycol-bis(β-aminoethyl ether)-N,N,N’,N’-tetraacetic acidis extensively used in molecular biology and protein biochemistry applications that require the chelation of divalent metal cations, such as calcium ions. This specificity makes it particularly useful in biological systems where calcium plays an important role, as well as for the determination of calcium in the presence of magnesium, since EGTA has greater specificity for calcium ions.