Learn More
Bioworld EGTA Buffer 0.5M, pH 8.0, 100ml
Catalog No. 50255957
Encompass_Preferred
This is a metal chelating agent specific for calcium ions and is commonly used in tissue culture media and biological buffers. Specifications: Sterile solution. pH : 8, Appearance Form : Liquid, Appearance Color : Clear, Melting Point : 241 °C. 100ml
Catalog No. 50-255-957 Supplier Bioworld Supplier No. 405200082
This is a metal chelating agent specific for calcium ions and is commonly used in tissue culture media and biological buffers. Specifications: Sterile solution. pH : 8, Appearance Form : Liquid, Appearance Color : Clear, Melting Point : 241 °C. 100ml