Learn More
Bioworld EGTA 1M, pH 8.0, EGTA 1M, pH 8.0 , 50ml
Catalog No. NC1280093
Encompass_Preferred
EGTA is a metal chelating agent specific for calcium ion that is commonly used reagent in tissue culture medias and biological buffers. Specifications: pH: 8.0, Sterile solution. Molecular Weight : 380.35 g/mol, Molecular Formula : C14H24N2O10, pH : 380.35, Appearance Form : Liquid, Appearance Color : clear. 50ml
Catalog No. NC1280093 Supplier Bioworld Supplier No. 401201281
EGTA is a metal chelating agent specific for calcium ion that is commonly used reagent in tissue culture medias and biological buffers. Specifications: pH: 8.0, Sterile solution. Molecular Weight : 380.35 g/mol, Molecular Formula : C14H24N2O10, pH : 380.35, Appearance Form : Liquid, Appearance Color : clear. 50ml