Learn More
Gold Biotechnology Inc EGTA 10 g
SDP
Catalog No. 5000562086
Encompass_Preferred
Ethyleneglycol bis(2-aminoethyl ether)-N N N N tetraacetic acid (EGTA) is a calcium specific chelator Because of its low affinity for Mg EGTA inhibits metalloproteases
Catalog No. 50-005-62086 Supplier Gold Biotechnology Inc Supplier No. E21710
Ethyleneglycol bis(2-aminoethyl ether)-N N N N tetraacetic acid (EGTA) is a calcium specific chelator Because of its low affinity for Mg EGTA inhibits metalloproteases