Learn More
Selleck Chemical LLC EGTA S4598-100mg
SDP
Catalog No. 503110265
Encompass_Preferred
EGTA is a specific calcium ion chelator This product has poor solubility animal experiments are available cell experiments please choose carefully
Catalog No. 50-311-0265 Supplier Selleck Chemical LLC Supplier No. S4598100MG
EGTA is a specific calcium ion chelator This product has poor solubility animal experiments are available cell experiments please choose carefully